Search
Search

MLO10| Gene Symbol: MLO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345729.1 | NP_001331804.1 | 12 - 13 | 1624 | 91 | |
| NM_125994.2 | NP_201398.1 | 13 - 14 | 1609 | 91 | ||
| GenBank mRNA | AF369571.1 | - | 13 - 14 | 1294 | 91 | |
| BT033095.1 | - | 13 - 14 | 1249 | 91 | ||
Set Membership: |
|