Search
Search

BGLU1| Gene Symbol: BGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333247.1 | NP_001323171.1 | 8 - 9 | 1144 | 103 | |
| NM_001333248.1 | NP_001323172.1 | 8 - 9 | 1095 | 103 | ||
| NM_001333249.1 | NP_001323173.1 | 8 - 9 | 862 | 103 | ||
| NM_001333250.1 | NP_001323174.1 | 8 - 9 | 862 | 103 | ||
| NM_179440.2 | NP_849771.2 | 9 - 10 | 898 | 103 | ||
| NM_202245.2 | NP_973974.2 | 8 - 9 | 862 | 103 | ||
Set Membership: |
|