Search
Search

ADT1| Gene Symbol: ADT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035947.1 | NP_001031024.1 | 9 - 10 | 1438 | 128 | |
| NM_101051.3 | NP_172644.1 | 10 - 11 | 1447 | 128 | ||
| GenBank mRNA | DQ411466.1 | - | 10 - 11 | 1128 | 128 | |
Set Membership: |
|