Search
Search

ADT1| Gene Symbol: ADT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035947.1 | NP_001031024.1 | 3 - 4 | 654 | 132 | |
| NM_101051.3 | NP_172644.1 | 3 - 4 | 749 | 132 | ||
| GenBank mRNA | DQ411466.1 | - | 3 - 4 | 430 | 132 | |
Set Membership: |
|