Search
Search

ACR8| Gene Symbol: ACR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332035.1 | NP_001321345.1 | 5 - 6 | 999 | 63 | |
| NM_101114.4 | NP_172704.1 | 6 - 7 | 1256 | 63 | ||
| GenBank mRNA | AF528064.1 | - | 6 - 7 | 1000 | 63 | |
| AK228687.1 | - | 6 - 7 | 1173 | 63 | ||
| BT026518.1 | - | 6 - 7 | 962 | 63 | ||
Set Membership: |
|