Search
Search

GLE1| Gene Symbol: GLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101182.4 | NP_172771.1 | 11 - 12 | 1891 | 72 | |
| GenBank mRNA | AK228913.1 | - | 11 - 12 | 1785 | 72 | |
| AX765852.1 | - | 10 - 11 | 1347 | 72 | ||
Set Membership: |
|