Search
Search

APK2A| Gene Symbol: APK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101304.4 | NP_172889.1 | 3 - 4 | 925 | 107 | |
| GenBank mRNA | AF334731.1 | - | 3 - 4 | 557 | 107 | |
| AY059894.1 | - | 3 - 4 | 749 | 107 | ||
| D88206.1 | - | 3 - 4 | 733 | 107 | ||
Set Membership: |
|