Search
Search

FBP7| Gene Symbol: FBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332527.1 | NP_001320240.1 | 3 - 4 | 524 | 112 | |
| NM_102025.3 | NP_564150.1 | 3 - 4 | 336 | 112 | ||
| GenBank mRNA | AK228033.1 | - | 3 - 4 | 256 | 112 | |
| BT006427.1 | - | 2 - 3 | 206 | 112 | ||
Set Membership: |
|