Search
Search

ERDL6| Gene Symbol: ERDL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106178.4 | NP_177658.1 | 10 - 11 | 1386 | 127 | |
| GenBank mRNA | AF412060.1 | - | 10 - 11 | 869 | 127 | |
| AY124845.1 | - | 10 - 11 | 858 | 127 | ||
Set Membership: |
|