Search
Search

GH9B7| Gene Symbol: GH9B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106219.3 | NP_177697.1 | 4 - 5 | 1486 | 74 | |
| GenBank mRNA | AY039938.1 | - | 4 - 5 | 1382 | 74 | |
| AY150451.1 | - | 4 - 5 | 1082 | 74 | ||
Set Membership: |
|