Search
Search

AMY2| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334710.1 | NP_001323338.1 | 5 - 6 | 663 | 80 | |
| NM_106262.4 | NP_177740.1 | 5 - 6 | 298 | 80 | ||
| GenBank mRNA | AK221564.1 | - | 5 - 6 | 298 | 80 | |
| AY084871.1 | - | 5 - 6 | 294 | 80 | ||
| BT025560.1 | - | 4 - 5 | 211 | 80 | ||
Set Membership: |
|