Search
Search

LACS9| Gene Symbol: LACS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334788.1 | NP_001321320.1 | 9 - 10 | 2048 | 140 | |
| NM_106407.7 | NP_177882.1 | 10 - 11 | 1975 | 140 | ||
| GenBank mRNA | AF503759.1 | - | 9 - 10 | 1700 | 140 | |
Set Membership: |
|