Search
Search

AT1G77600| Gene Symbol: AT1G77600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198491.1 | NP_001185420.1 | 7 - 8 | 1574 | 130 | |
| NM_001198492.1 | NP_001185421.1 | 7 - 8 | 1574 | 130 | ||
| NM_001334789.1 | NP_001319396.1 | 7 - 8 | 1511 | 130 | ||
Set Membership: |
|