Search
Search

TPPB| Gene Symbol: TPPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334818.1 | NP_001322316.1 | 5 - 6 | 716 | 87 | |
| NM_106458.4 | NP_177932.1 | 5 - 6 | 802 | 87 | ||
| GenBank mRNA | BT002801.1 | - | 5 - 6 | 711 | 87 | |
| BT004348.1 | - | 5 - 6 | 650 | 87 | ||
Set Membership: |
|