Search
Search

TPS1| Gene Symbol: TPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334832.1 | NP_001319403.1 | 4 - 5 | 2497 | 107 | |
| NM_001334833.1 | NP_001322931.1 | 3 - 4 | 2456 | 107 | ||
| NM_001334834.1 | NP_001322932.1 | 3 - 4 | 2423 | 107 | ||
| NM_106505.5 | NP_177979.1 | 3 - 4 | 687 | 107 | ||
| GenBank mRNA | KJ138926.1 | - | 2 - 3 | 481 | 107 | |
Set Membership: |
|