Search
Search

ALDH5F1| Gene Symbol: ALDH5F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106592.5 | NP_178062.1 | 18 - 19 | 1612 | 67 | |
| GenBank mRNA | AF117335.1 | - | 18 - 19 | 1530 | 67 | |
| AF428367.1 | - | 18 - 19 | 1532 | 67 | ||
| AY056147.1 | - | 18 - 19 | 1518 | 67 | ||
Set Membership: |
|