Search
Search

TED3| Gene Symbol: TED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106630.3 | NP_565222.1 | 7 - 8 | 1086 | 70 | |
| NM_202454.2 | NP_974183.1 | 7 - 8 | 1090 | 70 | ||
| GenBank mRNA | AY064063.1 | - | 7 - 8 | 1087 | 70 | |
| AY096373.1 | - | 7 - 8 | 886 | 70 | ||
Set Membership: |
|