Search
Search

RAE1| Gene Symbol: RAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106715.5 | NP_178182.1 | 9 - 10 | 1123 | 78 | |
| GenBank mRNA | AY087683.1 | - | 9 - 10 | 938 | 78 | |
| BT025241.1 | - | 8 - 9 | 845 | 78 | ||
Set Membership: |
|