Search
Search

ORP3B| Gene Symbol: ORP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111764.4 | NP_187541.1 | 9 - 10 | 1356 | 65 | |
| GenBank mRNA | AY062703.1 | - | 9 - 10 | 1321 | 65 | |
| AY093373.1 | - | 9 - 10 | 1135 | 65 | ||
Set Membership: |
|