Search
Search

MDHAR| Gene Symbol: MDHAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125135.1 | NP_001118607.1 | 3 - 4 | 221 | 83 | |
| NM_111829.4 | NP_566361.1 | 3 - 4 | 317 | 83 | ||
| GenBank mRNA | AY084556.1 | - | 3 - 4 | 219 | 83 | |
| AY093765.1 | - | 3 - 4 | 221 | 83 | ||
| BT001054.1 | - | 3 - 4 | 176 | 83 | ||
Set Membership: |
|