Search
Search

OMR1| Gene Symbol: OMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111840.3 | NP_187616.1 | 6 - 7 | 1696 | 102 | |
| GenBank mRNA | AF096281.1 | - | 6 - 7 | 1389 | 102 | |
| AY065037.1 | - | 6 - 7 | 1408 | 102 | ||
Set Membership: |
|