Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111851.6 | NP_187627.3 | 6 - 7 | 942 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |