Search
Search

AT3G10200| Gene Symbol: AT3G10200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111855.4 | NP_187631.2 | 6 - 7 | 1753 | 112 | |
| GenBank mRNA | AK228597.1 | - | 6 - 7 | 1617 | 112 | |
| BT005741.1 | - | 6 - 7 | 1617 | 112 | ||
Set Membership: |
|