Search
Search

SEC8| Gene Symbol: SEC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337871.1 | NP_001327644.1 | 24 - 25 | 3203 | 114 | |
| NM_111873.4 | NP_566372.1 | 24 - 25 | 3283 | 114 | ||
| GenBank mRNA | AK118541.1 | - | 24 - 25 | 3107 | 114 | |
| AY059763.1 | - | 24 - 25 | 3107 | 114 | ||
Set Membership: |
|