Search
Search

HB2| Gene Symbol: HB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111887.3 | NP_187663.1 | 2 - 3 | 377 | 74 | |
| GenBank mRNA | AY087650.1 | - | 2 - 3 | 307 | 74 | |
| BT028909.1 | - | 2 - 3 | 212 | 74 | ||
Set Membership: |
|