Search
Search

ASD1| Gene Symbol: ASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337892.1 | NP_001326565.1 | 16 - 17 | 2544 | 109 | |
| NM_001337893.1 | NP_001326566.1 | 16 - 17 | 2693 | 109 | ||
| NM_001337894.1 | NP_001326567.1 | 15 - 16 | 1818 | 109 | ||
| NM_111911.5 | NP_187685.1 | 16 - 17 | 2356 | 109 | ||
Set Membership: |
|