Search
Search

LBD25| Gene Symbol: LBD25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113681.2 | NP_566823.1 | 1 - 2 | 295 | 115 | |
| GenBank mRNA | AB164304.1 | - | 1 - 2 | 78 | 115 | |
| AB473836.1 | - | 1 - 2 | 254 | 115 | ||
| AF447892.1 | - | 1 - 2 | 247 | 115 | ||
Set Membership: |
|