Search
Search

CARA| Gene Symbol: CARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113690.3 | NP_566824.1 | 5 - 6 | 627 | 81 | |
| GenBank mRNA | AY046004.1 | - | 5 - 6 | 558 | 81 | |
| AY079315.1 | - | 5 - 6 | 484 | 81 | ||
Set Membership: |
|