Search
Search

CSLC04| Gene Symbol: CSLC04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113737.3 | NP_566835.1 | 3 - 4 | 1753 | 161 | |
| GenBank mRNA | AY056286.1 | - | 3 - 4 | 1635 | 161 | |
| AY091433.1 | - | 3 - 4 | 1354 | 161 | ||
| KJ138809.1 | - | 3 - 4 | 1354 | 161 | ||
Set Membership: |
|