Search
Search

PHS1| Gene Symbol: PHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113857.3 | NP_189578.1 | 1 - 2 | 300 | 67 | |
| GenBank mRNA | AY049235.1 | - | 1 - 2 | 285 | 67 | |
| BT003015.1 | - | 1 - 2 | 236 | 67 | ||
| KJ138807.1 | - | 1 - 2 | 236 | 67 | ||
Set Membership: |
|