Search
Search

UTR5| Gene Symbol: UTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339235.1 | NP_001326359.1 | 6 - 7 | 711 | 140 | |
| NM_114487.4 | NP_190204.2 | 6 - 7 | 761 | 140 | ||
| GenBank mRNA | BT011224.1 | - | 6 - 7 | 702 | 140 | |
| BT012152.1 | - | 6 - 7 | 676 | 140 | ||
Set Membership: |
|