Search
Search

TOC75-III| Gene Symbol: TOC75-III | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114541.3 | NP_190258.1 | 5 - 6 | 2078 | 110 | |
| GenBank mRNA | AY127014.1 | - | 5 - 6 | 2010 | 110 | |
| BT006358.1 | - | 5 - 6 | 1963 | 110 | ||
Set Membership: |
|