Search
Search

SELT| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114598.3 | NP_190314.2 | 3 - 4 | 597 | 75 | |
| GenBank mRNA | AK175455.1 | - | 3 - 4 | 566 | 75 | |
| AK176362.1 | - | 3 - 4 | 592 | 75 | ||
| BT010507.1 | - | 3 - 4 | 529 | 75 | ||
Set Membership: |
|