Search
Search

ATSMC2| Gene Symbol: ATSMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339310.1 | NP_001325474.1 | 13 - 14 | 2848 | 83 | |
| NM_114614.4 | NP_190330.1 | 13 - 14 | 2785 | 83 | ||
Set Membership: |
|