Search
Search

AT3G47660| Gene Symbol: AT3G47660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339324.1 | NP_001325561.1 | 6 - 7 | 1023 | 69 | |
| NM_001339325.1 | NP_001325560.1 | 6 - 7 | 749 | 69 | ||
| NM_001339327.1 | NP_001325562.1 | 5 - 6 | 576 | 69 | ||
| NM_114634.4 | NP_190350.4 | 6 - 7 | 499 | 69 | ||
Set Membership: |
|