Search
Search

LSU1| Gene Symbol: LSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203113.1 | NP_001190042.1 | 2 | 288 | 133 | |
| NM_114818.4 | NP_190527.1 | 1 | 501 | 133 | ||
| GenBank mRNA | AK117604.1 | - | 1 | 338 | 133 | |
| AY086614.1 | - | 1 | 345 | 133 | ||
Set Membership: |
|