Search
Search

UBP26| Gene Symbol: UBP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339429.1 | NP_001325635.1 | 4 - 5 | 1138 | 87 | |
| NM_001339430.1 | NP_001325634.1 | 4 - 5 | 1152 | 87 | ||
| NM_114820.4 | NP_566922.1 | 4 - 5 | 1249 | 87 | ||
Set Membership: |
|