Search
Search

GATA6| Gene Symbol: GATA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114968.4 | NP_190677.1 | 2 - 3 | 481 | 92 | |
| GenBank mRNA | AY064048.1 | - | 2 - 3 | 311 | 92 | |
| AY117315.1 | - | 1 - 2 | 260 | 92 | ||
Set Membership: |
|