Search
Search

CCS52A2| Gene Symbol: CCS52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117262.3 | NP_192929.2 | 5 - 6 | 1309 | 89 | |
| GenBank mRNA | AY063875.1 | - | 5 - 6 | 1264 | 89 | |
| AY091235.1 | - | 5 - 6 | 1139 | 89 | ||
Set Membership: |
|