Search
Search

AT4G19045| Gene Symbol: AT4G19045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160781.1 | NP_001154253.1 | 2 - 3 | 184 | 113 | |
| GenBank mRNA | AK117341.1 | - | 2 - 3 | 180 | 113 | |
| BT006214.1 | - | 2 - 3 | 72 | 113 | ||
Set Membership: |
|