Search
Search

DCL4| Gene Symbol: DCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203419.2 | NP_001190348.1 | 11 - 12 | 1843 | 87 | |
| NM_001343652.1 | NP_001330078.1 | 8 - 9 | 1011 | 87 | ||
| NM_122039.5 | NP_197532.3 | 11 - 12 | 1844 | 87 | ||
| GenBank mRNA | DQ118423.1 | - | 11 - 12 | 1844 | 87 | |
Set Membership: |
|