Search
Search

COPT5| Gene Symbol: COPT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122072.2 | NP_197565.1 | 1 | 538 | 97 | |
| GenBank mRNA | AY039578.1 | - | 1 | 477 | 97 | |
| AY086915.1 | - | 1 | 490 | 97 | ||
Set Membership: |
|