Search
Search

MS3| Gene Symbol: MS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085142.1 | NP_001078611.1 | 11 - 12 | 2076 | 124 | |
| NM_122107.4 | NP_197598.2 | 11 - 12 | 2129 | 124 | ||
| GenBank mRNA | AK229291.1 | - | 11 - 12 | 2076 | 124 | |
Set Membership: |
|