Search
Search

AT5G21060| Gene Symbol: AT5G21060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036839.2 | NP_001031916.1 | 3 - 4 | 462 | 112 | |
| NM_001036840.2 | NP_001031917.1 | 3 - 4 | 462 | 112 | ||
| NM_122114.3 | NP_197605.1 | 3 - 4 | 502 | 112 | ||
| GenBank mRNA | BT004002.1 | - | 3 - 4 | 371 | 112 | |
| BT005655.1 | - | 3 - 4 | 325 | 112 | ||
Set Membership: |
|