Search
Search

J2| Gene Symbol: J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122127.3 | NP_568412.1 | 1 - 2 | 354 | 129 | |
| GenBank mRNA | AK118386.1 | - | 1 - 2 | 227 | 129 | |
| BT026479.1 | - | 1 - 2 | 152 | 129 | ||
Set Membership: |
|