Search
Search

SGS3| Gene Symbol: SGS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343817.1 | NP_001331491.1 | 3 - 4 | 2218 | 94 | |
| NM_122263.3 | NP_197747.1 | 4 - 5 | 1812 | 94 | ||
| GenBank mRNA | BT002944.1 | - | 4 - 5 | 1596 | 94 | |
| BT004380.1 | - | 3 - 4 | 1505 | 94 | ||
Set Membership: |
|