Search
Search

LCB2| Gene Symbol: LCB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036855.1 | NP_001031932.1 | 1 - 2 | 355 | 68 | |
| NM_122272.3 | NP_197756.1 | 1 - 2 | 371 | 68 | ||
| GenBank mRNA | AY050829.1 | - | 1 - 2 | 318 | 68 | |
| AY133827.1 | - | 1 - 2 | 123 | 68 | ||
Set Membership: |
|