Search
Search

FRO4| Gene Symbol: FRO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122303.3 | NP_197786.2 | 1 - 2 | 206 | 79 | |
| GenBank mRNA | AF462813.1 | - | 1 - 2 | 199 | 79 | |
| BT010474.1 | - | 1 - 2 | 158 | 79 | ||
Set Membership: |
|