Search
Search

CYCA2;1| Gene Symbol: CYCA2;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343923.1 | NP_001318646.1 | 5 - 6 | 864 | 133 | |
| NM_001343924.1 | NP_001332474.1 | 5 - 6 | 864 | 133 | ||
| NM_001343925.1 | NP_001332473.1 | 4 - 5 | 666 | 133 | ||
| NM_122447.3 | NP_197920.2 | 4 - 5 | 648 | 133 | ||
Set Membership: |
|