Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYCA2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343923.1 | NP_001318646.1 | 6 - 7 | 992 | 66 | |
NM_001343924.1 | NP_001332474.1 | 6 - 7 | 992 | 66 | ||
NM_001343925.1 | NP_001332473.1 | 5 - 6 | 794 | 66 | ||
NM_122447.3 | NP_197920.2 | 5 - 6 | 776 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |